The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020151.3 | NP_064536.2 | 6 - 7 | 1242 | 102 |
XM_011511499.1 | XP_011509801.1 | 6 - 7 | 859 | 102 | |
GenBank mRNA | AF270647.1 | - | 6 - 7 | 1169 | 102 |
AK023234.1 | - | 5 - 6 | 501 | 102 | |
AK054615.1 | - | 6 - 7 | 608 | 102 | |
AK096666.1 | - | 2 - 3 | 154 | 102 | |
AK291190.1 | - | 6 - 7 | 624 | 102 | |
BC003028.1 | - | 6 - 7 | 1225 | 102 | |
BC007894.2 | - | 7 - 8 | 1000 | 102 | |
BC008894.2 | - | 6 - 7 | 1225 | 102 | |
BC009998.2 | - | 7 - 8 | 1000 | 102 | |
BC012774.2 | - | 6 - 7 | 1192 | 102 | |
BC012793.2 | - | 6 - 7 | 1206 | 102 | |
BC013279.2 | - | 6 - 7 | 1225 | 102 | |
BC014076.2 | - | 6 - 7 | 1202 | 102 | |
BC014274.2 | - | 6 - 7 | 1221 | 102 | |
BC032106.1 | - | 6 - 7 | 1216 | 102 |