The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020158.3 | NP_064543.3 | 1 - 2 | 173 | 127 |
GenBank mRNA | AF281134.1 | - | 1 - 2 | 173 | 127 |
AF285785.1 | - | 1 - 2 | 304 | 127 | |
AK314941.1 | - | 1 - 2 | 158 | 127 | |
BC007742.2 | - | 1 - 2 | 156 | 127 | |
BC107696.1 | - | 1 - 2 | 178 | 127 |