The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_028288.1 | 6 - 7 | 1666 | 84 | |
GenBank mRNA | AB035332.1 | - | 3 - 4 | 3445 | 84 |
AB035334.1 | - | 4 - 5 | 2477 | 84 | |
AB035335.1 | - | 3 - 4 | 2022 | 84 | |
AB035337.1 | - | 6 - 7 | 1668 | 84 | |
AB035338.1 | - | 6 - 7 | 1863 | 84 | |
AB035339.1 | - | 1 - 2 | 134 | 84 | |
AF195821.1 | - | 2 - 3 | 268 | 84 |