Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
JPH3
Entrez Gene ID
57338
Gene Name
junctophilin 3
Gene Aliases
CAGL237, HDL2, JP-3, JP3, TNRC22
Chromosome Location
Chr.16: 87601835 - 87698156 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020655.3 NP_065706.2 4 - 5 2431 73
NR_073379.2 4 - 5 1885 73
GenBank mRNA AB042636.1 - 4 - 5 2340 73
AK055486.1 - 3 - 4 1242 73
AK126663.1 - 3 - 4 2465 73
AK295518.1 - 4 - 5 1885 73
AK297215.1 - 1 - 2 401 73
BC036533.2 - 4 - 5 2413 73

View Full Product Details