Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRX
Entrez Gene ID
57716
Gene Name
periaxin
Gene Aliases
CMT4F
Chromosome Location
Chr.19: 40393762 - 40414718 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020956.2 NP_066007.1 5 - 6 452 104
NM_181882.2 NP_870998.2 5 - 6 452 104
XM_011527171.2 XP_011525473.1 5 - 6 444 104
GenBank mRNA AB046840.1 - 1 - 2 582 104
AF321191.1 - 5 - 6 452 104
AF321192.1 - 5 - 6 452 104
BC067266.1 - 5 - 6 454 104

View Full Product Details