The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040142.1 | NP_001035232.1 | 11 - 12 | 1961 | 76 |
NM_001040143.1 | NP_001035233.1 | 10 - 11 | 1722 | 76 | |
NM_021007.2 | NP_066287.2 | 11 - 12 | 2027 | 76 | |
XM_005246753.3 | XP_005246810.1 | 11 - 12 | 1977 | 76 | |
XM_005246755.4 | XP_005246812.1 | 6 - 7 | 1062 | 76 | |
XM_017004655.1 | XP_016860144.1 | 11 - 12 | 1876 | 76 | |
XM_017004656.1 | XP_016860145.1 | 11 - 12 | 1932 | 76 | |
XM_017004657.1 | XP_016860146.1 | 11 - 12 | 1829 | 76 | |
XM_017004658.1 | XP_016860147.1 | 11 - 12 | 2141 | 76 | |
GenBank mRNA | AB208888.1 | - | 11 - 12 | 1814 | 76 |
BP311214.1 | - | 2 - 3 | 419 | 76 | |
M94055.1 | - | 11 - 12 | 1776 | 76 |