The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021146.3 | NP_066969.1 | 1 - 2 | 672 | 61 |
XM_017000004.1 | XP_016855493.1 | 1 - 2 | 637 | 61 | |
GenBank mRNA | AB593026.1 | - | 1 - 2 | 606 | 61 |
AB593027.1 | - | 1 - 2 | 673 | 61 | |
AK313716.1 | - | 1 - 2 | 638 | 61 | |
AY358301.1 | - | 1 - 2 | 595 | 61 | |
BC001881.1 | - | 1 - 2 | 620 | 61 | |
BT009802.1 | - | 1 - 2 | 381 | 61 | |
Y16132.1 | - | 1 - 2 | 388 | 61 |