The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021175.3 | NP_066998.1 | 1 - 2 | 324 | 111 |
GenBank mRNA | AF309489.1 | - | 1 - 2 | 163 | 111 |
AJ277280.1 | - | 1 - 2 | 130 | 111 | |
AY358669.1 | - | 1 - 2 | 132 | 111 | |
BC020612.1 | - | 1 - 2 | 142 | 111 | |
BP306149.1 | - | 1 - 2 | 326 | 111 |