Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSKS
Entrez Gene ID
60385
Gene Name
testis specific serine kinase substrate
Gene Aliases
PPP1R161, STK22S1, TSKS1, TSSKS
Chromosome Location
Chr.19: 49739753 - 49763328 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021733.1 NP_068379.1 2 - 3 410 60
XM_011527201.1 XP_011525503.1 2 - 3 480 60
XM_011527202.1 XP_011525504.1 2 - 3 480 60
XM_011527203.2 XP_011525505.1 2 - 3 480 60
GenBank mRNA BC058862.1 - 2 - 3 438 60

View Full Product Details