The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001126121.1 | NP_001119593.1 | 7 - 8 | 998 | 72 |
NM_001126122.1 | NP_001119594.1 | 6 - 7 | 908 | 72 | |
NM_021734.4 | NP_068380.3 | 7 - 8 | 982 | 72 | |
XM_005257559.3 | XP_005257616.1 | 7 - 8 | 1620 | 72 | |
XM_005257560.2 | XP_005257617.1 | 7 - 8 | 1174 | 72 | |
XM_005257561.3 | XP_005257618.1 | 7 - 8 | 1510 | 72 | |
XM_005257562.2 | XP_005257619.1 | 7 - 8 | 1157 | 72 | |
XM_006722007.2 | XP_006722070.1 | 7 - 8 | 1030 | 72 | |
XM_017024926.1 | XP_016880415.1 | 6 - 7 | 1572 | 72 | |
XM_017024927.1 | XP_016880416.1 | 5 - 6 | 1246 | 72 | |
GenBank mRNA | AF182404.1 | - | 5 - 6 | 774 | 72 |
AJ251857.1 | - | 5 - 6 | 812 | 72 | |
AK075239.1 | - | 8 - 9 | 1004 | 72 | |
AK097882.1 | - | 5 - 6 | 1684 | 72 | |
AK225881.1 | - | 5 - 6 | 811 | 72 | |
AL833387.1 | - | 7 - 8 | 949 | 72 | |
AY346372.1 | - | 4 - 5 | 603 | 72 | |
BC001075.2 | - | 7 - 8 | 910 | 72 | |
BC005120.1 | - | 7 - 8 | 910 | 72 | |
BM455504.1 | - | 4 - 5 | 516 | 72 |