The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301130.1 | NP_001288059.1 | 2 - 3 | 221 | 104 |
NM_001301131.1 | NP_001288060.1 | 2 - 3 | 221 | 104 | |
NM_021974.4 | NP_068809.1 | 2 - 3 | 221 | 104 | |
XM_017028832.1 | XP_016884321.1 | 2 - 3 | 187 | 104 | |
GenBank mRNA | BC003582.1 | - | 2 - 3 | 199 | 104 |
BP195499.1 | - | 2 - 3 | 187 | 104 | |
CR456546.1 | - | 2 - 3 | 111 | 104 | |
CR457021.1 | - | 2 - 3 | 96 | 104 | |
DA372532.1 | - | 2 - 3 | 221 | 104 | |
DN989996.1 | - | 2 - 3 | 196 | 104 |