Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FXYD7
Entrez Gene ID
53822
Gene Name
FXYD domain containing ion transport regulator 7
Gene Aliases
-
Chromosome Location
Chr.19: 35143250 - 35154302 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022006.1 NP_071289.1 1 - 2 116 120
GenBank mRNA AI929519.1 - 1 - 2 116 120
AK057825.1 - 1 - 2 91 120
BC018619.1 - 1 - 2 100 120

View Full Product Details