Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GPR88
Entrez Gene ID
54112
Gene Name
G protein-coupled receptor 88
Gene Aliases
COCPMR, STRG
Chromosome Location
Chr.1: 100538172 - 100542027 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022049.2 NP_071332.2 1 - 2 333 64
GenBank mRNA AB042410.1 - 1 - 2 366 64
BC036775.1 - 1 - 2 333 64

View Full Product Details