The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022066.3 | NP_071349.3 | 9 - 10 | 1696 | 99 |
XM_005257567.1 | XP_005257624.1 | 9 - 10 | 1680 | 99 | |
XM_017024934.1 | XP_016880423.1 | 9 - 10 | 1605 | 99 | |
XM_017024935.1 | XP_016880424.1 | 9 - 10 | 1605 | 99 | |
GenBank mRNA | AB051521.1 | - | 9 - 10 | 1696 | 99 |
AK025999.1 | - | 1 - 2 | 172 | 99 | |
AK225412.1 | - | 1 - 2 | 172 | 99 | |
AL832432.1 | - | 2 - 3 | 621 | 99 | |
BC036820.1 | - | 4 - 5 | 832 | 99 |