The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022131.2 | NP_071414.2 | 12 - 13 | 2231 | 113 |
XM_017007022.1 | XP_016862511.1 | 12 - 13 | 2272 | 113 | |
GenBank mRNA | AJ278018.1 | - | 13 - 14 | 2051 | 113 |
AK315308.1 | - | 13 - 14 | 2231 | 113 | |
AY753303.1 | - | 13 - 14 | 2041 | 113 | |
BC103496.1 | - | 13 - 14 | 2051 | 113 | |
BC103506.1 | - | 12 - 13 | 2050 | 113 | |
BC104485.1 | - | 13 - 14 | 2048 | 113 |