Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POPDC3
Entrez Gene ID
64208
Gene Name
popeye domain containing 3
Gene Aliases
POP3, bA355M14.1
Chromosome Location
Chr.6: 105157900 - 105179983 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022361.4 NP_071756.2 2 - 3 891 66
XM_011536067.2 XP_011534369.1 2 - 3 1051 66
XM_017011194.1 XP_016866683.1 2 - 3 2334 66
XM_017011195.1 XP_016866684.1 2 - 3 1794 66
GenBank mRNA AF204171.1 - 1 - 2 636 66
AK314101.1 - 2 - 3 891 66
BC022323.2 - 1 - 2 741 66

View Full Product Details