The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022373.4 | NP_071768.3 | 6 - 7 | 1276 | 70 |
XM_006715765.1 | XP_006715828.1 | 4 - 5 | 1084 | 70 | |
XM_011515473.1 | XP_011513775.1 | 3 - 4 | 1282 | 70 | |
XM_017012506.1 | XP_016867995.1 | 5 - 6 | 1474 | 70 | |
GenBank mRNA | AK025966.1 | - | 1 - 2 | 132 | 70 |
AK055708.1 | - | 5 - 6 | 1423 | 70 | |
AK315685.1 | - | 5 - 6 | 900 | 70 | |
BC005091.1 | - | 6 - 7 | 1082 | 70 | |
BC020264.1 | - | 6 - 7 | 1233 | 70 |