Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NXN
Entrez Gene ID
64359
Gene Name
nucleoredoxin
Gene Aliases
NRX, TRG-4
Chromosome Location
Chr.17: 799313 - 979766 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205319.1 NP_001192248.1 7 - 8 864 65
NM_022463.4 NP_071908.2 7 - 8 1207 65
XM_005256756.4 XP_005256813.1 6 - 7 1033 65
XM_005256758.3 XP_005256815.1 7 - 8 1536 65
GenBank mRNA AF086523.1 - 1 - 2 35 65
AK022676.1 - 5 - 6 636 65
AK027451.1 - 6 - 7 760 65
AK297990.1 - 3 - 4 336 65
AK302073.1 - 7 - 8 866 65
AY189690.1 - 6 - 7 747 65
BC009327.2 - 7 - 8 801 65
BC063828.1 - 4 - 5 462 65
BC073845.1 - 5 - 6 617 65

View Full Product Details