Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HOXC8
Entrez Gene ID
3224
Gene Name
homeobox C8
Gene Aliases
HOX3, HOX3A
Chromosome Location
Chr.12: 54009106 - 54012763 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022658.3 NP_073149.1 1 - 2 620 80
GenBank mRNA AK290959.1 - 1 - 2 611 80
BC053898.1 - 1 - 2 620 80

View Full Product Details