The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001257994.1 | NP_001244923.1 | 7 - 8 | 1625 | 91 |
NM_022727.5 | NP_073564.3 | 7 - 8 | 1625 | 91 | |
NM_182984.4 | NP_892029.2 | 8 - 9 | 1501 | 91 | |
XM_011530139.2 | XP_011528441.1 | 8 - 9 | 1432 | 91 | |
XM_011530141.1 | XP_011528443.1 | 6 - 7 | 807 | 91 | |
XM_011530142.2 | XP_011528444.1 | 4 - 5 | 465 | 91 | |
XM_017028770.1 | XP_016884259.1 | 7 - 8 | 1635 | 91 | |
GenBank mRNA | AK025106.1 | - | 6 - 7 | 1021 | 91 |
AK057029.1 | - | 7 - 8 | 1575 | 91 | |
BC013352.2 | - | 7 - 8 | 1270 | 91 | |
BC017184.2 | - | 8 - 9 | 1387 | 91 | |
BC108251.1 | - | 7 - 8 | 1305 | 91 | |
CR456354.1 | - | 7 - 8 | 1346 | 91 |