Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IQCH
Entrez Gene ID
64799
Gene Name
IQ motif containing H
Gene Aliases
NYDSP5
Chromosome Location
Chr.15: 67254772 - 67501804 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031715.2 NP_001026885.1 6 - 7 733 64
NM_001284348.1 NP_001271277.1 3 - 4 399 64
NM_001284349.1 NP_001271278.1 3 - 4 399 64
NM_001322470.1 NP_001309399.1 3 - 4 399 64
NM_001322473.1 NP_001309402.1 3 - 4 399 64
NM_001322474.1 NP_001309403.1 3 - 4 399 64
NM_001322475.1 NP_001309404.1 4 - 5 517 64
XM_011521907.2 XP_011520209.1 7 - 8 874 64
XM_011521908.2 XP_011520210.1 6 - 7 733 64
XM_011521911.2 XP_011520213.1 5 - 6 661 64
XM_011521912.2 XP_011520214.1 7 - 8 874 64
XM_011521913.2 XP_011520215.1 7 - 8 875 64
XM_011521914.2 XP_011520216.1 4 - 5 393 64
XM_011521915.2 XP_011520217.1 7 - 8 874 64
XM_011521916.2 XP_011520218.1 3 - 4 312 64
XM_011521917.2 XP_011520219.1 5 - 6 581 64
XM_011521918.2 XP_011520220.1 3 - 4 367 64
XM_011521922.2 XP_011520224.1 7 - 8 877 64
XM_017022491.1 XP_016877980.1 7 - 8 874 64
XM_017022492.1 XP_016877981.1 7 - 8 874 64
XM_017022493.1 XP_016877982.1 7 - 8 874 64
XM_017022494.1 XP_016877983.1 7 - 8 874 64
XM_017022495.1 XP_016877984.1 7 - 8 874 64
XM_017022496.1 XP_016877985.1 7 - 8 874 64
XM_017022497.1 XP_016877986.1 7 - 8 874 64
XM_017022498.1 XP_016877987.1 7 - 8 874 64
XM_017022499.1 XP_016877988.1 7 - 8 874 64
XM_017022500.1 XP_016877989.1 4 - 5 540 64
XM_017022501.1 XP_016877990.1 3 - 4 367 64
XM_017022502.1 XP_016877991.1 3 - 4 422 64
GenBank mRNA AK022538.1 - 3 - 4 349 64
AK292478.1 - 6 - 7 685 64
AK304303.1 - 6 - 7 681 64
AY014282.2 - 6 - 7 704 64
BC031041.1 - 3 - 4 351 64
BC048964.1 - 6 - 7 683 64

View Full Product Details