The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282167.1 | NP_001269096.1 | 12 - 13 | 1581 | 84 |
NM_022825.3 | NP_073736.2 | 12 - 13 | 1436 | 84 | |
NM_203473.2 | NP_982299.1 | 13 - 14 | 1454 | 84 | |
NM_203474.1 | NP_982300.1 | 12 - 13 | 1306 | 84 | |
NM_203475.2 | NP_982301.1 | 14 - 15 | 1469 | 84 | |
XM_005272635.2 | XP_005272692.1 | 13 - 14 | 1809 | 84 | |
XM_005272636.2 | XP_005272693.1 | 12 - 13 | 1791 | 84 | |
XM_005272637.1 | XP_005272694.1 | 15 - 16 | 1751 | 84 | |
XM_006724545.3 | XP_006724608.1 | 15 - 16 | 1534 | 84 | |
XM_006724546.3 | XP_006724609.1 | 15 - 16 | 1606 | 84 | |
XM_006724547.1 | XP_006724610.1 | 14 - 15 | 1326 | 84 | |
XM_011543948.2 | XP_011542250.1 | 12 - 13 | 1720 | 84 | |
XM_017029735.1 | XP_016885224.1 | 14 - 15 | 1834 | 84 | |
XM_017029736.1 | XP_016885225.1 | 12 - 13 | 1293 | 84 | |
GenBank mRNA | AF317058.1 | - | 13 - 14 | 1290 | 84 |
AF317059.1 | - | 12 - 13 | 1275 | 84 | |
AF317060.1 | - | 12 - 13 | 1272 | 84 | |
AF317061.1 | - | 11 - 12 | 1257 | 84 | |
AK056340.1 | - | 12 - 13 | 1763 | 84 | |
AK057727.1 | - | 11 - 12 | 1683 | 84 | |
AK293347.1 | - | 12 - 13 | 1546 | 84 | |
AK314745.1 | - | 13 - 14 | 1339 | 84 | |
AK316378.1 | - | 11 - 12 | 1328 | 84 | |
BC019080.2 | - | 13 - 14 | 1401 | 84 | |
L08239.1 | - | 12 - 13 | 1206 | 84 |