The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024091.3 | NP_076996.2 | 3 - 4 | 1697 | 93 |
NR_036553.1 | 2 - 3 | 146 | 93 | ||
NR_073608.1 | 2 - 3 | 146 | 93 | ||
XM_006714498.1 | XP_006714561.1 | 3 - 4 | 1608 | 93 | |
GenBank mRNA | AF161354.1 | - | 2 - 3 | 120 | 93 |
AK026927.1 | - | 3 - 4 | 1662 | 93 | |
BC001295.1 | - | 2 - 3 | 1189 | 93 | |
BC113563.1 | - | 2 - 3 | 1612 | 93 | |
BC143517.1 | - | 2 - 3 | 1612 | 93 | |
BC143518.1 | - | 3 - 4 | 1049 | 93 | |
BG613096.1 | - | 2 - 3 | 136 | 93 |