The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042467.2 | NP_001035932.1 | 2 - 3 | 353 | 95 |
NM_001281473.1 | NP_001268402.1 | 2 - 3 | 353 | 95 | |
NM_001281474.1 | NP_001268403.1 | 3 - 4 | 503 | 95 | |
NM_024101.6 | NP_077006.1 | 2 - 3 | 353 | 95 | |
NR_104019.1 | 2 - 3 | 353 | 95 | ||
XM_006712737.1 | XP_006712800.1 | 2 - 3 | 321 | 95 | |
XM_006712739.1 | XP_006712802.1 | 2 - 3 | 321 | 95 | |
XM_006712740.1 | XP_006712803.1 | 2 - 3 | 321 | 95 | |
XM_017004893.1 | XP_016860382.1 | 2 - 3 | 321 | 95 | |
XM_017004894.1 | XP_016860383.1 | 2 - 3 | 321 | 95 | |
GenBank mRNA | AK022207.1 | - | 2 - 3 | 321 | 95 |
AK094168.1 | - | 3 - 4 | 503 | 95 | |
AK225381.1 | - | 2 - 3 | 298 | 95 | |
AK292929.1 | - | 2 - 3 | 321 | 95 | |
AK296745.1 | - | 2 - 3 | 322 | 95 | |
AK310821.1 | - | 2 - 3 | 321 | 95 | |
BC001653.2 | - | 2 - 3 | 285 | 95 | |
BC051269.1 | - | 2 - 3 | 295 | 95 | |
BM272410.1 | - | 2 - 3 | 347 | 95 | |
DC300186.1 | - | 2 - 3 | 353 | 95 |