The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195731.1 | NP_001182660.1 | 3 - 4 | 938 | 80 |
NM_024536.5 | NP_078812.2 | 3 - 4 | 1294 | 80 | |
GenBank mRNA | AB086063.1 | - | 3 - 4 | 1128 | 80 |
AB095813.1 | - | 3 - 4 | 1303 | 80 | |
AK302124.1 | - | 3 - 4 | 938 | 80 | |
AL136814.1 | - | 2 - 3 | 441 | 80 | |
AY358403.1 | - | 3 - 4 | 1294 | 80 | |
BC021223.2 | - | 3 - 4 | 1294 | 80 | |
BC023531.2 | - | 3 - 4 | 1245 | 80 |