The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168468.1 | NP_001161940.1 | 4 - 5 | 368 | 105 |
NM_024791.3 | NP_079067.3 | 4 - 5 | 368 | 105 | |
NR_033122.1 | 4 - 5 | 444 | 105 | ||
XM_011543000.2 | XP_011541302.1 | 3 - 4 | 663 | 105 | |
XM_011543001.2 | XP_011541303.1 | 3 - 4 | 757 | 105 | |
XM_011543002.2 | XP_011541304.1 | 3 - 4 | 849 | 105 | |
XM_011543005.2 | XP_011541307.1 | 4 - 5 | 353 | 105 | |
XM_017018334.1 | XP_016873823.1 | 2 - 3 | 1077 | 105 | |
XM_017018335.1 | XP_016873824.1 | 2 - 3 | 1077 | 105 | |
XM_017018336.1 | XP_016873825.1 | 2 - 3 | 1077 | 105 | |
XM_017018337.1 | XP_016873826.1 | 2 - 3 | 1077 | 105 | |
GenBank mRNA | AB094096.1 | - | 2 - 3 | 1060 | 105 |
AK026409.1 | - | 4 - 5 | 350 | 105 | |
AK091966.1 | - | 3 - 4 | 663 | 105 | |
AK298333.1 | - | 4 - 5 | 426 | 105 | |
AK315403.1 | - | 4 - 5 | 345 | 105 | |
AY047359.1 | - | 4 - 5 | 330 | 105 | |
BC029042.1 | - | 4 - 5 | 339 | 105 | |
BX648391.1 | - | 4 - 5 | 427 | 105 |