Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CEP76
Entrez Gene ID
79959
Gene Name
centrosomal protein 76
Gene Aliases
C18orf9, HsT1705
Chromosome Location
Chr.18: 12661733 - 12702777 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271989.1 NP_001258918.1 5 - 6 813 79
NM_024899.3 NP_079175.2 6 - 7 1038 79
NR_073537.1 6 - 7 1033 79
XM_005258149.4 XP_005258206.1 6 - 7 1017 79
XM_017025981.1 XP_016881470.1 6 - 7 1017 79
XM_017025982.1 XP_016881471.1 6 - 7 1098 79
XM_017025984.1 XP_016881473.1 6 - 7 1000 79
XM_017025985.1 XP_016881474.1 6 - 7 966 79
XM_017025986.1 XP_016881475.1 5 - 6 775 79
XM_017025987.1 XP_016881476.1 5 - 6 742 79
GenBank mRNA AK022604.1 - 6 - 7 900 79
AK301964.1 - 5 - 6 775 79
AK303117.1 - 5 - 6 760 79
BC026307.1 - 6 - 7 965 79
BC033026.1 - 6 - 7 900 79

View Full Product Details