Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 15450834-15451329 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COLQ
Entrez Gene ID
8292
Gene Name
collagen like tail subunit of asymmetric acetylcholinesterase
Gene Aliases
CMS5, EAD
UniGene
Hs.146735
Gene Chromosome Location
Chr.3: 15450133-15521751 on Build GRCh38
Transcripts
NM_005677.3 NM_080538.2 NM_080539.3

SNP Detail

SNP ID Chromosome Location
rs1010891408 Chr.3: 15451092-15451092 on Build GRCh38
rs886058096 Chr.3: 15451219-15451219 on Build GRCh38
rs577687841 Chr.3: 15450940-15450940 on Build GRCh38
rs541202497 Chr.3: 15451153-15451153 on Build GRCh38
rs1052089682 Chr.3: 15451200-15451200 on Build GRCh38
rs878961057 Chr.3: 15451106-15451106 on Build GRCh38
rs907644682 Chr.3: 15450883-15450883 on Build GRCh38
rs2278962 Chr.3: 15451279-15451279 on Build GRCh38
rs564176947 Chr.3: 15451222-15451222 on Build GRCh38
rs973068700 Chr.3: 15450906-15450906 on Build GRCh38
rs916413795 Chr.3: 15451138-15451138 on Build GRCh38
rs2278961 Chr.3: 15451300-15451300 on Build GRCh38
rs771628214 Chr.3: 15451236-15451236 on Build GRCh38
rs932259649 Chr.3: 15450938-15450938 on Build GRCh38
rs537057072 Chr.3: 15450941-15450941 on Build GRCh38
rs763440701 Chr.3: 15451218-15451218 on Build GRCh38
rs372672661 Chr.3: 15451248-15451248 on Build GRCh38
rs544927968 Chr.3: 15451128-15451128 on Build GRCh38
rs1052489174 Chr.3: 15451062-15451062 on Build GRCh38
rs1014610240 Chr.3: 15451186-15451186 on Build GRCh38
rs892063592 Chr.3: 15450885-15450885 on Build GRCh38
rs921563059 Chr.3: 15451224-15451224 on Build GRCh38
rs983122079 Chr.3: 15450873-15450873 on Build GRCh38
rs1004703460 Chr.3: 15451176-15451176 on Build GRCh38
rs116231717 Chr.3: 15451152-15451152 on Build GRCh38
rs770219393 Chr.3: 15451180-15451180 on Build GRCh38
rs747056584 Chr.3: 15451061-15451061 on Build GRCh38
rs925034609 Chr.3: 15451164-15451164 on Build GRCh38
rs116450586 Chr.3: 15451089-15451089 on Build GRCh38
rs1045127509 Chr.3: 15451104-15451104 on Build GRCh38
rs557314953 Chr.3: 15451090-15451090 on Build GRCh38
rs576307598 Chr.3: 15451094-15451094 on Build GRCh38
rs569980572 Chr.3: 15451237-15451237 on Build GRCh38
rs922188126 Chr.3: 15450929-15450929 on Build GRCh38
rs934958863 Chr.3: 15451187-15451187 on Build GRCh38
rs746360529 Chr.3: 15451140-15451140 on Build GRCh38
rs534228650 Chr.3: 15451231-15451231 on Build GRCh38
rs368981773 Chr.3: 15450893-15450893 on Build GRCh38
rs1035613813 Chr.3: 15451137-15451137 on Build GRCh38
rs999197428 Chr.3: 15450872-15450873 on Build GRCh38
rs149076823 Chr.3: 15451048-15451049 on Build GRCh38
rs533015053 Chr.3: 15451223-15451223 on Build GRCh38
rs10154896 Chr.3: 15451129-15451129 on Build GRCh38
rs952019644 Chr.3: 15451085-15451085 on Build GRCh38
rs1052031317 Chr.3: 15450939-15450939 on Build GRCh38
rs935533456 Chr.3: 15450984-15450984 on Build GRCh38
rs912009121 Chr.3: 15450949-15450949 on Build GRCh38

View Full Product Details