The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025099.5 | NP_079375.3 | 11 - 12 | 2004 | 87 |
NR_046431.1 | 11 - 12 | 1899 | 87 | ||
XM_006721577.3 | XP_006721640.1 | 10 - 11 | 1859 | 87 | |
XM_006721578.3 | XP_006721641.1 | 11 - 12 | 1988 | 87 | |
XM_011524010.2 | XP_011522312.1 | 11 - 12 | 1883 | 87 | |
XM_011524011.2 | XP_011522313.1 | 10 - 11 | 1753 | 87 | |
GenBank mRNA | AK025823.1 | - | 1 - 2 | 147 | 87 |
AK091077.1 | - | 11 - 12 | 1955 | 87 | |
AK225375.1 | - | 1 - 2 | 147 | 87 | |
AL831955.1 | - | 11 - 12 | 1846 | 87 | |
BC026057.2 | - | 2 - 3 | 145 | 87 | |
BC110373.1 | - | 3 - 4 | 371 | 87 | |
BC111783.1 | - | 11 - 12 | 1955 | 87 | |
DQ451688.1 | - | 11 - 12 | 1945 | 87 | |
FJ790670.1 | - | 11 - 12 | 1945 | 87 |