The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042400.1 | NP_001035859.1 | 1 - 2 | 151 | 83 |
XM_005247799.3 | XP_005247856.1 | 1 - 2 | 150 | 83 | |
XM_011513194.2 | XP_011511496.1 | 1 - 2 | 149 | 83 | |
XM_017007247.1 | XP_016862736.1 | 1 - 2 | 149 | 83 | |
XM_017007259.1 | XP_016862748.1 | 1 - 2 | 149 | 83 | |
XM_017007262.1 | XP_016862751.1 | 1 - 2 | 149 | 83 | |
XM_017007266.1 | XP_016862755.1 | 1 - 2 | 149 | 83 | |
GenBank mRNA | AK023448.1 | - | 1 - 2 | 97 | 83 |
BC068997.1 | - | 1 - 2 | 71 | 83 |