Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZBP1
Entrez Gene ID
81030
Gene Name
Z-DNA binding protein 1
Gene Aliases
C20orf183, DAI, DLM-1, DLM1
Chromosome Location
Chr.20: 57603846 - 57620576 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160418.1 NP_001153890.1 3 - 4 558 82
NM_001160419.2 NP_001153891.1 4 - 5 783 82
NM_001323966.1 NP_001310895.1 3 - 4 558 82
NM_030776.2 NP_110403.2 4 - 5 783 82
NR_136660.1 3 - 4 558 82
XM_011529057.2 XP_011527359.1 4 - 5 633 82
XM_011529058.2 XP_011527360.1 4 - 5 633 82
GenBank mRNA AJ300575.1 - 4 - 5 680 82
AK122761.1 - 3 - 4 561 82
AK128405.1 - 2 - 3 2743 82
AK290918.1 - 4 - 5 633 82
AX775973.1 - 4 - 5 680 82
AX775975.1 - 4 - 5 680 82
BC028218.1 - 3 - 4 1297 82
BC131706.1 - 4 - 5 598 82
CT002036.1 - 4 - 5 601 82

View Full Product Details