Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C18orf21
Entrez Gene ID
83608
Gene Name
chromosome 18 open reading frame 21
Gene Aliases
HsT3108, PNAS-124, PNAS-131, XTP13
Chromosome Location
Chr.18: 35972625 - 35979287 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001201476.1 NP_001188405.1 2 - 3 204 77
NM_031446.4 NP_113634.3 2 - 3 204 77
GenBank mRNA AF277182.1 - 2 - 3 63 77
AF277189.1 - 2 - 3 63 77
BC108730.1 - 2 - 3 164 77
BC119765.1 - 2 - 3 155 77
BC119766.1 - 2 - 3 154 77
BX460890.2 - 2 - 3 530 77
DN991579.1 - 2 - 3 127 77

View Full Product Details