The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300761.2 | NP_001287690.1 | 7 - 8 | 921 | 63 |
NM_001317806.1 | NP_001304735.1 | 6 - 7 | 820 | 63 | |
NM_031956.3 | NP_114162.2 | 6 - 7 | 820 | 63 | |
NR_133922.1 | 6 - 7 | 820 | 63 | ||
XM_005263270.1 | XP_005263327.1 | 7 - 8 | 903 | 63 | |
XM_006714335.1 | XP_006714398.1 | 7 - 8 | 903 | 63 | |
XM_006714336.1 | XP_006714399.1 | 7 - 8 | 903 | 63 | |
XM_011532310.2 | XP_011530612.1 | 7 - 8 | 903 | 63 | |
GenBank mRNA | AF345910.1 | - | 6 - 7 | 771 | 63 |
AK093145.1 | - | 7 - 8 | 867 | 63 | |
AK225725.1 | - | 6 - 7 | 767 | 63 | |
AK301807.1 | - | 6 - 7 | 746 | 63 | |
AK314066.1 | - | 6 - 7 | 816 | 63 | |
BC024193.2 | - | 6 - 7 | 762 | 63 | |
EF432564.1 | - | 4 - 5 | 589 | 63 |