Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPNS1
Entrez Gene ID
83985
Gene Name
sphingolipid transporter 1 (putative)
Gene Aliases
HSpin1, LAT, PP2030, SPIN1, SPINL, nrs
Chromosome Location
Chr.16: 28974767 - 28984769 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142448.1 NP_001135920.1 3 - 4 452 69
NM_001142450.1 NP_001135922.1 3 - 4 602 69
NM_001142451.1 NP_001135923.1 2 - 3 689 69
NM_032038.2 NP_114427.1 2 - 3 689 69
XM_006721096.3 XP_006721159.1 3 - 4 459 69
XM_017023757.1 XP_016879246.1 3 - 4 460 69
XM_017023758.1 XP_016879247.1 3 - 4 456 69
XM_017023759.1 XP_016879248.1 3 - 4 455 69
GenBank mRNA AF212371.1 - 2 - 3 636 69
AF370423.1 - 3 - 4 584 69
AK095677.1 - 2 - 3 686 69
AK289787.1 - 2 - 3 684 69
AL524908.3 - 3 - 4 577 69
BC006156.1 - 2 - 3 300 69
BC008325.1 - 2 - 3 514 69
BC038961.1 - 3 - 4 466 69
BC047741.1 - 2 - 3 663 69
BC065235.1 - 3 - 4 434 69
BG771918.1 - 2 - 3 697 69

View Full Product Details