The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002049.3 | NP_002040.1 | 1 - 2 | 73 | 99 |
XM_011543897.2 | XP_011542199.1 | 1 - 2 | 107 | 99 | |
GenBank mRNA | BC009797.1 | - | 1 - 2 | 76 | 99 |
X17254.1 | - | 1 - 2 | 94 | 99 |