The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320415.1 | NP_001307344.1 | 3 - 4 | 377 | 61 |
NM_002382.4 | NP_002373.3 | 3 - 4 | 377 | 61 | |
NM_145112.2 | NP_660087.1 | 2 - 3 | 350 | 61 | |
NM_145113.2 | NP_660088.1 | 3 - 4 | 377 | 61 | |
NR_073137.1 | 2 - 3 | 296 | 61 | ||
XM_011536773.2 | XP_011535075.1 | 3 - 4 | 365 | 61 | |
XM_017021312.1 | XP_016876801.1 | 2 - 3 | 336 | 61 | |
XM_017021313.1 | XP_016876802.1 | 3 - 4 | 256 | 61 | |
GenBank mRNA | AK290130.1 | - | 2 - 3 | 281 | 61 |
AK290929.1 | - | 3 - 4 | 350 | 61 | |
AK292189.1 | - | 3 - 4 | 287 | 61 | |
AK292630.1 | - | 3 - 4 | 225 | 61 | |
AL833643.1 | - | 3 - 4 | 353 | 61 | |
AY353088.1 | - | 2 - 3 | 145 | 61 | |
BC003525.1 | - | 2 - 3 | 260 | 61 | |
BC004516.2 | - | 3 - 4 | 303 | 61 | |
BC025685.1 | - | 2 - 3 | 297 | 61 | |
BC027924.1 | - | 2 - 3 | 307 | 61 | |
BC036092.1 | - | 3 - 4 | 323 | 61 | |
BG177865.1 | - | - | 60 | ||
CR542191.1 | - | 2 - 3 | 145 | 61 | |
DB451269.1 | - | 2 - 3 | 298 | 61 | |
M64240.1 | - | 3 - 4 | 199 | 61 | |
X60287.1 | - | 3 - 4 | 193 | 61 |