The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005515.3 | NP_005506.3 | 1 - 2 | 995 | 71 |
GenBank mRNA | AF107457.1 | - | 1 - 2 | 879 | 71 |
X56537.1 | - | 1 - 2 | 202 | 71 |