The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005924.4 | NP_005915.2 | 1 - 2 | 801 | 71 |
GenBank mRNA | AK309472.1 | - | 3 - 4 | 430 | 71 |
AK313386.1 | - | 1 - 2 | 726 | 71 | |
BC017021.1 | - | 1 - 2 | 698 | 71 | |
CR536553.1 | - | 1 - 2 | 517 | 71 | |
L36328.1 | - | 1 - 2 | 546 | 71 | |
X82629.1 | - | 1 - 2 | 782 | 71 |