The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007348.3 | NP_031374.2 | 6 - 7 | 810 | 95 |
XM_006711224.1 | XP_006711287.1 | 6 - 7 | 734 | 95 | |
XM_011509308.1 | XP_011507610.1 | 6 - 7 | 734 | 95 | |
XM_011509309.1 | XP_011507611.1 | 6 - 7 | 734 | 95 | |
XM_011509310.2 | XP_011507612.1 | 6 - 7 | 734 | 95 | |
GenBank mRNA | AB015856.1 | - | 6 - 7 | 761 | 95 |
AB208929.1 | - | 6 - 7 | 698 | 95 | |
AF005887.1 | - | 6 - 7 | 735 | 95 |