Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MMP10
Entrez Gene ID
4319
Gene Name
matrix metallopeptidase 10
Gene Aliases
SL-2, STMY2
Chromosome Location
Chr.11: 102770502 - 102780628 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002425.2 NP_002416.1 9 - 10 1370 82
GenBank mRNA AK222601.1 - 9 - 10 1370 82
AK313960.1 - 9 - 10 1370 82
BC002591.2 - 9 - 10 1357 82
BT007442.1 - 9 - 10 1333 82
X07820.1 - 9 - 10 1355 82

View Full Product Details