The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006022.3 | NP_006013.1 | 1 - 2 | 419 | 67 |
GenBank mRNA | AJ222700.1 | - | 1 - 2 | 314 | 67 |
AK312345.1 | - | 1 - 2 | 353 | 67 | |
BC000456.2 | - | 1 - 2 | 335 | 67 | |
BC016867.1 | - | 1 - 2 | 338 | 67 | |
BC069207.1 | - | 1 - 2 | 327 | 67 | |
BC105277.1 | - | 1 - 2 | 330 | 67 | |
BC146664.1 | - | 1 - 2 | 334 | 67 | |
BC146679.1 | - | 1 - 2 | 334 | 67 | |
BG703493.1 | - | 1 - 2 | 425 | 67 | |
CR456711.1 | - | 1 - 2 | 123 | 67 | |
D38585.1 | - | 1 - 2 | 123 | 67 | |
D87061.1 | - | 1 - 2 | 123 | 67 | |
U35048.1 | - | 1 - 2 | 305 | 67 |