The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136017.3 | NP_001129489.1 | 4 - 5 | 908 | 67 |
NM_001136125.2 | NP_001129597.1 | 3 - 4 | 695 | 67 | |
NM_001136126.2 | NP_001129598.1 | 3 - 4 | 692 | 67 | |
NM_001287427.1 | NP_001274356.1 | 4 - 5 | 1411 | 67 | |
NM_001287434.1 | NP_001274363.1 | 4 - 5 | 874 | 67 | |
NM_001760.4 | NP_001751.1 | 4 - 5 | 911 | 67 | |
GenBank mRNA | AB209825.1 | - | 4 - 5 | 702 | 67 |
AK055519.1 | - | 4 - 5 | 969 | 67 | |
AK057206.1 | - | 4 - 5 | 702 | 67 | |
AK096276.1 | - | 4 - 5 | 1222 | 67 | |
AK097856.1 | - | 3 - 4 | 486 | 67 | |
AK315421.1 | - | 4 - 5 | 827 | 67 | |
BC011616.2 | - | 4 - 5 | 793 | 67 | |
BX117734.1 | - | 1 - 2 | 121 | 67 | |
CR542246.1 | - | 4 - 5 | 712 | 67 | |
DA069566.1 | - | 3 - 4 | 511 | 67 | |
M90814.1 | - | 4 - 5 | 812 | 67 | |
M92287.1 | - | 4 - 5 | 877 | 67 |