The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000425.4 | NP_000416.1 | 4 - 5 | 615 | 102 |
NM_001143963.2 | NP_001137435.1 | 3 - 4 | 499 | 102 | |
NM_001278116.1 | NP_001265045.1 | 5 - 6 | 624 | 102 | |
NM_024003.3 | NP_076493.1 | 4 - 5 | 514 | 102 | |
GenBank mRNA | AB102653.1 | - | 4 - 5 | 400 | 102 |
AK289754.1 | - | 4 - 5 | 431 | 102 | |
BC126229.1 | - | 4 - 5 | 516 | 102 | |
BC136447.1 | - | 4 - 5 | 516 | 102 | |
EF506611.1 | - | 3 - 4 | 391 | 102 | |
M74387.1 | - | 4 - 5 | 417 | 102 | |
M77640.1 | - | 4 - 5 | 426 | 102 | |
X59847.1 | - | 4 - 5 | 406 | 102 |