The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000755.3 | NP_000746.2 | 1 - 2 | 306 | 76 |
GenBank mRNA | AL523652.3 | - | 1 - 2 | 298 | 76 |
BC000723.2 | - | 1 - 2 | 245 | 76 | |
BI752361.1 | - | 1 - 2 | 299 | 76 | |
BQ068230.1 | - | - | 330 | 76 |