The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002481.3 | NP_002472.2 | 10 - 11 | 1607 | 150 |
XM_005245206.4 | XP_005245263.1 | 10 - 11 | 1607 | 150 | |
XM_017001342.1 | XP_016856831.1 | 10 - 11 | 1607 | 150 | |
XM_017001343.1 | XP_016856832.1 | 10 - 11 | 1607 | 150 | |
XM_017001344.1 | XP_016856833.1 | 10 - 11 | 1607 | 150 | |
XM_017001345.1 | XP_016856834.1 | 10 - 11 | 1607 | 150 | |
XM_017001346.1 | XP_016856835.1 | 10 - 11 | 1607 | 150 | |
XM_017001347.1 | XP_016856836.1 | 10 - 11 | 1607 | 150 | |
XM_017001348.1 | XP_016856837.1 | 10 - 11 | 1607 | 150 | |
XM_017001349.1 | XP_016856838.1 | 10 - 11 | 1607 | 150 | |
XM_017001350.1 | XP_016856839.1 | 11 - 12 | 1664 | 150 | |
XM_017001351.1 | XP_016856840.1 | 10 - 11 | 1607 | 150 | |
GenBank mRNA | AB003062.1 | - | 10 - 11 | 1620 | 150 |
AF324888.1 | - | 10 - 11 | 1601 | 150 | |
AK307591.1 | - | 10 - 11 | 1607 | 150 | |
BC144699.1 | - | 10 - 11 | 1599 | 150 |