Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ILF3
Entrez Gene ID
3609
Gene Name
interleukin enhancer binding factor 3
Gene Aliases
CBTF, DRBF, DRBP76, MMP4, MPHOSPH4, MPP4, NF-AT-90, NF110, NF110b, NF90, NF90a, NF90b, NFAR, NFAR-1, NFAR2, TCP110, TCP80
Chromosome Location
Chr.19: 10654261 - 10692419 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001137673.1 NP_001131145.1 1 - 2 221 106
NM_004516.3 NP_004507.2 1 - 2 221 106
NM_012218.3 NP_036350.2 1 - 2 221 106
NM_017620.2 NP_060090.2 1 - 2 221 106
NM_153464.2 NP_703194.1 1 - 2 221 106
XM_006722742.2 XP_006722805.1 1 - 2 136 106
XM_011527985.2 XP_011526287.1 1 - 2 136 106
XM_017026765.1 XP_016882254.1 1 - 2 136 106
GenBank mRNA AB209062.1 - 1 - 2 178 106
AK291205.1 - 1 - 2 120 106
AK291617.1 - 1 - 2 106 106
AK293170.1 - 1 - 2 125 106
AK294084.1 - 1 - 2 112 106
BC001770.1 - 1 - 2 106 106
BC064836.1 - 1 - 2 154 106
DC320902.1 - 1 - 2 224 106
U10324.1 - 1 - 2 168 106
X98264.1 - 1 - 2 108 106

View Full Product Details