The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006686.3 | NP_006677.1 | 1 - 1 | 232 | 56 |
GenBank mRNA | AK313871.1 | - | 1 - 1 | 225 | 56 |
BC033789.1 | - | 1 - 1 | 210 | 56 | |
DB021848.1 | - | 1 - 1 | 269 | 56 |