The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271815.1 | NP_001258744.1 | 10 - 11 | 1538 | 98 |
NM_007052.4 | NP_008983.2 | 11 - 12 | 1649 | 98 | |
XM_017029407.1 | XP_016884896.1 | 9 - 10 | 1298 | 98 | |
GenBank mRNA | AF127763.2 | - | - | 1649 | 98 |
AF166327.1 | - | 11 - 12 | 1513 | 98 | |
AK292201.1 | - | 11 - 12 | 1604 | 98 | |
AK313678.1 | - | 11 - 12 | 1558 | 98 | |
BC075014.2 | - | 11 - 12 | 1464 | 98 | |
BC075015.2 | - | 11 - 12 | 1464 | 98 | |
DQ424904.1 | - | 10 - 11 | 1332 | 98 |