Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PCDHGA8
Entrez Gene ID
9708
Gene Name
protocadherin gamma subfamily A, 8
Gene Aliases
PCDH-GAMMA-A8
Chromosome Location
Chr.5: 141391916 - 141512979 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014004.2 NP_054723.1 1 - 1 876 78
NM_032088.1 NP_114477.1 1 - 1 876 78
GenBank mRNA AB002325.1 - 1 - 1 2637 78
BC152415.1 - 1 - 1 2637 78

View Full Product Details