The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130703.1 | NP_001124175.1 | 3 - 4 | 411 | 68 |
NM_001130705.1 | NP_001124177.1 | 2 - 3 | 305 | 68 | |
GenBank mRNA | AF134708.1 | - | 1 - 2 | 200 | 68 |
DB039551.1 | - | 2 - 3 | 260 | 68 | |
DB051105.1 | - | 3 - 4 | 397 | 68 | |
DB073142.1 | - | 3 - 4 | 414 | 68 |